AccuForge® GEM® Single Edge Blade Cartridges, Uncoated
Supplier: Accutec Blades
AGBL-7051-0000 AGBL-7051-BULK AGBL-7052-0000 AGBL-7011-0000 AGBL-7012-0000 AGBL-7014-0000
77322-510CS 783.25 CAD
77322-510 77322-512 77322-514 77322-470 77322-472 77322-478
AccuForge® GEM® Single Edge Blade Cartridges, Uncoated
Razors and Blades
AccuForge® Single Edge blades feature precision ground edges for sharpness and durability.
  • Paper-free packaging is cleanroom ready
  • Choose 3-facet blades for increased sharpness, or 2-facet blades for increased durability
  • Individually packed in clamshell with product key and instructions for use
  • Available in bulk pack without clamshells
  • Available in 0.012" heavy-duty carbon steel
  • Each cartridge contains 90 to 100 blades
Use with GEM RUNNER™ Dispenser for improved safety and productivity.
Caution: Blades are extremely sharp. Wear eye protection.More Product Information