AccuForge® GEM® Single Edge Blade Cartridge, Coated
Supplier: Accutec Blades
AGBL-7013-0000 AGBL-7013-BULK
77322-474CS 1373.32 CAD
77322-474 77322-476
AccuForge® GEM® Single Edge Blade Cartridge, Coated
Razors and Blades
AccuForge® Single Edge blades feature 3-facet edges for sharpness. The proprietary MicroCoat® Enrichment Technology reduces cut force and increases durability.
  • Degreased blades for reduced oil and particulates
  • Cartridge inserts into reloadable safety dispenser
  • Paper-free packaging is cleanroom ready
  • Individually packed in clamshell with product key and instructions for use
  • Available in bulk pack without clamshells
  • Each cartridge contains 100 blades
  • Made in USA
MicroCoat® blade enrichment technology adds a layer of protection for each blade, helping maintain a sharp and effective edge over multiple uses.
Use with GEM RUNNER™ Dispenser for improved safety and productivity.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.