AccuForge® Double Edge Blades, Unwrapped
Supplier: Accutec Blades
AGBL-6002-0000 AGBL-6005-0000
77322-446CS 15693.37 CAD
77322-446 77322-450
AccuForge® Double Edge Blades, Unwrapped
Razors and Blades
AccuForge® Double Edge blades feature a 3-Facet edge for exacting, clean cuts. The proprietary MicroCoat® Enrichment Technology reduces cut force and helps the blade maintain a sharp, effective edge over multiple uses.
  • Blade edge with MicroCoat® technology enhances durability and sharpness
  • 3-Facet edge for increased sharpness
  • Available degreased for reduced oil and particulates
  • Blades are supplied on a bayonet
  • Bulk unwrapped blades are ideal for high volume uses
  • Made in USA
MicroCoat® blade enrichment technology adds a layer of protection for each blade, helping maintain a sharp and effective edge over multiple uses.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.