AccuForge® Hobby Blade #11, Stainless Steel
Supplier: AccuTec Blades
AccuForge®
AGBL-2001-0000
77322-444CS 1050.19 CAD
77322-444
AccuForge® Hobby Blade #11, Stainless Steel
Razors and Blades
AccuForge® #11 Hobby blades are constructed from stainless steel for increased sharpness and corrosion resistance. Precision acute point for fine cuts.
  • Fits all standard #1 Handles
  • High-quality stainless steel blade
  • Bulk pack of 1000 blades
  • Made in USA
Excellent for precision cutting and trimming.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.