Eppendorf® 5425/5425 R Rotors for Centrifuge
Supplier: Eppendorf
5495500006 5495503005 5495505008 5495508007 5495510001 5495512004
CA76204-612EA 1562.44 CAD
CA76204-612 CA76204-614 CA76204-616 CA76204-624 CA76204-626 CA76204-628
Eppendorf® 5425/5425 R Rotors for Centrifuge
Centrifuges Rotors
Eppendorf rotors for centrifuge 5425/5425 R, precision meets versatility. Optimize your centrifuge with a range of capacities and speeds. Explore accessory options such as adaptors and buckets for tailored sample processing, ensuring optimal results in molecular biology and beyond.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 minutes)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
  • Aerosol-tight and PTFE coated options available
Refine your centrifugation experience with Eppendorf rotors designed for centrifuge 5425/5425 R. Offering versatility in capacities and speeds, these rotors cater to a spectrum of applications, from microcentrifugation to larger sample volumes. Accessories such as adaptors and buckets further customize your setup for specific needs. Achieve reliable sample separation with precision and flexibility, meeting the demands of molecular biology, genomics, and beyond. Explore the comprehensive range to optimize your centrifuge for reproducible results.