Lab Line 3950V and 3970V Vertical Autoclaves: Autoclave everything in your laboratory, including liquids. Designed with simplicity and economy in mind.
- Plug & play – no plumbing required and no need to connect to building utilities
- Built-in condensation reservoir
- Designed to autoclave tall bioreactors
- Two integrated temperature probes for complete sterilization of liquids
- 3 year-warranty
The 3950 and 3970V lab autoclaves were developed specifically for education institutions and research facilities. Designed with simplicity and economy in mind. These vertical floor-standing autoclaves cover the fundamental needs for general laboratory sterilization, including two integrated temperature probes to ensure complete sterilization of your liquid samples, with the aim of increasing the productivity of your laboratory. Offering high capacity, together with the optimization of resources such as water, power and operating time. An affordable and reliable solution you can count on for managing your laboratory workload.
Certifications: CE marked
Ordering information: Stainless steel wire baskets and containers available for all autoclave models (sold separately).
Delivery information: Units includes 2 large stainless steel wire baskets, funnel, drain tubing, a bottom base plate, and 1 buck boost transformer