Rotors for Centrifuges, 5810/5810 R
Supplier: Eppendorf
022638602 022638807 022638009 022638041 5820740000 5820755008 5820754001 022638611
CA47743-060EA 7588.33 CAD
CA47743-060 CA47745-376 CA53513-866 CA53513-872 CA11017-374 CA11017-378 CA11017-380 75875-114
Rotors for Centrifuges, 5810/5810 R
Centrifuges Rotors
Eppendorf Rotors for centrifuges 5810/5810 R - precision redefined. Tailor your high-capacity centrifugation with varied rotor options. Choose from a range of capacities and speeds, accessorize with versatile buckets, and ensure efficient sample processing. Ideal for diverse applications, guaranteeing accuracy and reproducibility.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 min)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
Redesign your high-capacity centrifugation with Eppendorf rotors crafted for centrifuges 5810/5810 R. These rotors offer a spectrum of capacities and speeds, providing adaptability for various sample processing needs. Accessorize with versatile buckets, ensuring efficient customization of your centrifuge setup. Ideal for diverse applications, the Centrifuge 5810 series is further enhanced with these precision-engineered rotors, guaranteeing accuracy and reproducibility in every spin. Explore the range to optimize your centrifugation experience.