Student Dissection Kits
Supplier: AFTAB AHMED SE
10-PMAK-1 10-PMDK-1 10-PMVK-1
470350-630CS 1570.85 CAD
470350-630 470350-632 470350-634
Student Dissection Kits
Dissection Sets
Pre-built instrument kits.
  • Three storage options
  • Easily store between uses
  • Quick setup for student groups
  • Easy instrument check when finished
These three dissection sets provide varied options for what instruments and storage best fit the class. Soft cases are lighter and a little easier to store.
The hard clamshell case provides the most protection from possible sliding instruments, while closing securely. Zippered cases provide storage for a greater variety of instruments, and when closed prevent the instruments from falling out if they weren't securely replaced.
Each set contains different instruments, see attached document for specific contents, with all containing scalpels, scissors, forceps, and teasing needles.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.