Hemostatic Forceps
Supplier: AFTAB AHMED SE
14-0551 20-105
470334-792BX 69.25 CAD
470334-792 470334-816
Hemostatic Forceps
Forceps
Lock for a secure hold.
  • Stainless steel
  • Serrated jaws
Hemostatic forceps are a locking forceps, with thumb loops that operate much like scissors. Often referred to as clamps, these have a serrated jaw to provide a secure hold while minimizing tissue damage.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.