Special Use Forceps
Supplier: AFTAB AHMED SE
14-0512 14-0590 14-0520 15-TW088 15-122 14-1010
470334-772BX 69.75 CAD
470334-772 470334-782 470334-784 470334-812 470334-814 470334-840
Special Use Forceps
Forceps
Specialty forceps for student use.
  • Stainless steel
  • A variety of options available
One set of forceps isn't like every other. This selection provides options for many users. Tweezers are a basic sharp point forceps with a very short blade length that makes the ends very easy to control and provides a stronger grip.
Cover slipping forceps are designed with a broader smooth tip that is bent at an angle to improve applying a coverslip to a microscope slide.
Microdissection forceps are similar to a watchmakers forceps with a lighter weight blade, requiring less force to close. Easy grip forceps have an enlarged grip loop part-way down each blade to provide additional stability and ease of closure for a narrow blade.
Finally the self closing forceps act in a reverse motion to most. Squeezing opens the jaws, and they close when released. These are easier to use if long-term holding is required.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.