Watchmaker Forceps
Supplier: AFTAB AHMED SE
14-1400 14-0505
470334-802BX 110.35 CAD
470334-802 470334-804
Watchmaker Forceps
Forceps
Ultra-fine point forceps.
  • Stainless steel
Extremely fine tips and heavier handles allow for the manipulation of very fine tissue samples with a secure hold.
These forceps have a smooth finish inside the jaws, to ensure a minimum of tissue damage.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.