Dissection Needles
Supplier: AFTAB AHMED SE
20-103 14-0633 14-1033
470334-820BX 93.7 CAD
470334-820 470334-822 470334-848
Dissection Needles
Dissection Needles
Sharp point needles.
  • Fine point teasing needles
  • Straight or angled tips
Very sharp points on these needles make tissue separation fast and easy. Used mostly to break apart muscle tissues.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.