Dissection Knives
Supplier: AFTAB AHMED SE
14-1190 14-0969 14-0930
470334-844BX 162.9 CAD
470334-844 470334-852 470334-890
Dissection Knives
Knives
Great for rough dissection.
  • Stainless steel construction
  • Fixed blades for safety
More durable than scalpels, dissection knives are great for rough dissection, rapid fleshing, or dealing with tough tissues. The basic fleshing knife has a shorter blade and is easily controlled.
The Amputation knife has an extended blade for long smooth cuts through soft tissues. Use the cartilage knife, which has a scalpel form, to easily reach into tight areas to cut tough tissue.
Caution: These instruments are intended for training and educational use only.More Product Information