Heavy-Duty 50 GSM SMS Autoclavable Bowl Covers with Steam Indicator
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
BWL-KCRP-300-20 BWL-KCRP-300-24 BWL-KCRP-300-30 BWL-KCRP-300-36 BWL-KCRP-300-40 BWL-KCRP-300-42 BWL-KCRP-300-48 BWL-KCRP-300-50 BWL-KCRP-300-54 BWL-KCRP-300-60
77395-072CS 4040.85 CAD
77395-072 77395-074 77395-076 77395-078 77395-080 77395-082 77395-084 77395-086 77395-088 77395-090
Heavy-Duty 50 GSM SMS Autoclavable Bowl Covers with Steam Indicator
Covers
This 50 g/m² SMS autoclavable bowl cover provides a microbial barrier that is ideal for pharmaceutical production applications.
  • Hydrophobic, tear-resistant, and puncture-resistant
  • Creates secondary level of protection during aseptic presentation and setup of equipment
  • Designed for autoclave cycles (121 to 135 °C)
  • Manufactured and packed in a certified ISO 8 cleanroom
  • Made in USA
Eliminate the need for cutting sheets of sterilization wrapping materials and securing them with autoclave tape and rubber bands. After the outer bag is removed, this stopper bowl cover should remain in place, providing protection from microbial and particulate contamination while the remaining set-up activities are performed. Used in conjunction with a secondary autoclave bag, it creates an effective wrapping system.
Certifications: Certificates of conformance are sent with every shipment.
Packaging: Cleanroom packed in sealed, labeled poly-bags with the lot number and date of manufacture. Covers are packed 1 per bag.