Blue Heavy-Duty Autoclavable SMS Muff Covers
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
MFKCRP300-03 MFKCRP300-04 MFKCRP300-05 MFKCRP300-06 MFKCRP300-07 MFKCRP300-08 MFKCRP300-09 MFKCRP300-12 MFKCRP300-16 MFKCRP300-18
77395-112CS 2346.3 CAD
77395-112 77395-114 77395-116 77395-118 77395-120 77395-122 77395-124 77395-126 77395-128 77395-130
Blue Heavy-Duty Autoclavable SMS Muff Covers
Covers
This 50 g/m² SMS autoclavable muff cover provides an excellent microbial barrier that is ideal for pharmaceutical production applications.
  • Eliminates autoclave tape residue
  • Hydrophobic, tear-resistant, and puncture-resistant
  • Creates secondary level of protection during aseptic presentation and setup of equipment
  • Designed for autoclave cycles (121 to 135 °C)
  • Manufactured and packed in a certified ISO 8 cleanroom
  • Used in conjunction with a secondary autoclave bag, it creates an effective wrapping system
  • Made in USA
Eliminate the need for cutting sheets of sterilization wrapping materials and securing them with autoclave tape and rubber bands. Cost-effective, time-saving approach standardizes the equipment preparation process, minimizes particulate generation, while also ensuring good aseptic technique upon removal after sterilization.
Certifications: Certificates of conformance are sent with every shipment.
Packaging: Cleanroom packed in sealed, labeled poly-bags with the lot number and date of manufacture. Covers are packed 100 per bag.