Eppendorf® Lab Shaker Clamps
Supplier: Eppendorf
ACE-1000S ACE-10S ACE-2000S ACE-3000S ACE-4000S ACE-5000S ACE-6000S ACFE-2800S ACSB-1000S ACSB-500S M1190-9000 M1190-9001 M1190-9003 M1190-9004 M1190-9005 M1190-9002 ACFE-2800S
CA89173-650EA 109.93 CAD
CA89173-650 CA89173-652 CA89173-654 CA89173-656 CA89173-658 CA89173-660 CA89173-662 CA89173-664 CA89173-666 CA89173-668 CA89173-706 CA89173-708 CA89173-712 CA89173-714 CA89173-716 CA89173-710 89173-664
Eppendorf® Lab Shaker Clamps
Shakers
Unlock the potential of your experiments with Eppendorf's versatile range of shaker clamps. Securely fasten Erlenmeyer flasks, Thomson Ultra Yield®, Optimum Growth™, Tunair®, Corning® and more to your universal platforms. Find the perfect clamp for every flask type and elevate your research today.
  • Stainless-steel construction (not spot welded or riveted) for added strength and safety
  • Clamps 2 L and above have an extra girdle for added security when agitating flasks at high speeds
  • Supplied complete with required screws, and are fitted with coiled wire springs for added stability at elevated speeds