epT.I.P.S.® Bulk - Pipette Tips
Supplier: Eppendorf
022492004 022492021 022492039 022492047 022492055 022492063 022492071 022492080 022492098 022492101 022494018 022492012 0030000650 0030000650
CA47745-164CS 142.79 CAD
CA47745-164 CA47745-168 CA47745-170 CA47745-172 CA47745-174 CA47745-176 CA47745-178 CA47745-180 CA47747-986 CA47747-992 CA89126-938 CA47745-166 75845-008 CA75845-008
epT.I.P.S.® Bulk - Pipette Tips
Pipette Tips
Eppendorf epT.I.P.S.® Bulk packs offer a practical, high-volume solution with 'Eppendorf Quality' pipette tips, ensuring precise and reliable measurements for various lab applications, packaged economically for high-throughput settings.
  • Original, high-quality Eppendorf pipette tips packaged in resealable bags
  • Available in sizes from 10 µl to 10 ml in Eppendorf Quality purity grade
  • epT.I.P.S.® Standard 200, 300 and 1000 µl tips are also available color-coded yellow and blue
  • epT.I.P.S.® Standard pipette tips can be autoclaved at 121 °C in 20 min
  • Fine
More Product Information