Mini Adapters, No. 7 Ace-Thred with PTFE Ferrule, Ace Glass Incorporated
Supplier: Ace Glass
5028-27 5028-31
89051-106EA 98.47 CAD
89051-106 89051-112
Mini Adapters, No. 7 Ace-Thred with PTFE Ferrule, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
These mini adapters are suitable for most vacuum work.
  • Inner joint at bottom
  • No. 7 Ace-Thred at top
  • Manufactured from borosilicate glass
Adapters feature an inner joint at bottom and No. 7 Ace-Thred at top.
Ordering information: Supplied with nylon bushing and PTFE ferrule in place of O-ring. For replacement ferrules, see 89075-734 series.