VWR® Flexible Bellow Joints, PTFE
Supplier: VWR International
89026-132 89026-134 89026-138
89026-132EA 426.62 CAD
89026-132 89026-134 89026-138
VWR® Flexible Bellow Joints, PTFE
Adapters Adapters, Conical Ground Joints
Flexible bellows are constructed of pure, non-contaminating PTFE.
  • Connect directly to standard ground glass joints
  • Usable up to 260°C (500°F)
  • Inert
Bellows correct misalignment and relieve stress in glassware assemblies. Transverse sealing rings provide medium to high vacuum performance and reduce the probability of glass bursting during temperature cycling.