Pocket Colorimeter II Test Kits
Supplier: Hach
2105528
2668000
2430000
2106069
2105569
2802246
2105669
1406499
2508025
2502025
2503025
2770900
2507025
2506025
2105628
1407099
2106169
2242000
2527025
CA21055-28PK
611.54
CAD
CA21055-28
CA2668000
CA58700-02
CA97009-434
CA97009-436
CA97009-438
CA97009-454
CA97009-456
CA97009-464
CA97009-470
CA97009-490
CA97009-494
CA97009-498
CA97009-500
CA97009-504
CA97009-512
CA97009-520
CA97009-536
CA97009-554
CA97009-560
CA97009-566
CA97009-468
CA97009-474
CA97009-488
CA97009-514
Pocket Colorimeter II Test Kits
Environmental Test Kits
This colorimeter is a portable, rugged, and convenient instrument, ideally suited for in-the-field analysis.
Tough, impact-resistant shell is also waterproof and floats. Four buttons allow easy operation. Accuracy is comparable to expensive lab instruments. All units are factory-programmed; no manual calibration is ever required. All units come with enough reagent sets for 100 tests.