AlphaSat® CleanPaks® Presaturated Wipers, Texwipe®
Supplier: ITW Texwipe
TX1034 TX1039 TX1034 TX1039
CATWTX1034CS 2478.86 CAD
CATWTX1034 CATWTX1039 TWTX1034 TWTX1039
AlphaSat® CleanPaks® Presaturated Wipers, Texwipe®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Wipers are made from 100% continuous-filament knit polyester and are presaturated with a blend of 70% 0.2 µm filtered semiconductor-grade isopropyl alcohol and 30% deionized water.
Note: Specifications reflect the data for the wiper in an unsaturated state.
The double-knit, no-run interlock pattern provides maximum abrasion resistance and low particle values. These wipers offer portion control and reduced potential of isopropyl alcohol spillage in and out of the cleanroom. Ideal for critical environments in medical device and integrated circuit manufacturing.
Ordering information: More Product Information