UltraSORB® Wipers, Foamtec International WCC
Supplier: Foamtec International
HT4644 HT4666 HT4699
10810-836CS 2220.15 CAD
10810-836 10810-842 10810-904
UltraSORB® Wipers, Foamtec International WCC
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Wipers are made from a fully cross-linked, naturally hydrophillic polyurethane foam and laundered with a proprietary process in an ISO Class 4 (FED-STD-209E Class 10/M2.5) environment.
  • UltraSORB® foam has a 3 lbs./ft³ density
  • Also offer outstanding wet tensile strength and abrasion resistance
Wipers are particle-trapping, abrasion-resistant, thermally stable, solvent-resistant, nonpyrogenic, and feature a sponge-like feel and softness, making them suitable for the wipedown of ISO Class 3 and Class 4 (FED-STD-209E Class 1/M1.5 and Class 10/M2.5) cleanrooms, optical surfaces, and hot and aseptic fill areas.
They are engineered specifically to minimize contaminants released from the wiper while maximizing the amount of particulates and residue removed from surfaces.