KIMBLE® KONTES® Distilling Adapters, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
169500-2440 275050-1420
KT169500-2440EA 313.96 CAD
KT169500-2440 KT275050-1420
KIMBLE® KONTES® Distilling Adapters, DWK Life Sciences
Distillation Components Distillation Adapters
Distilling adapter is useful in the sub-boiling separation of mixtures with volatile components.
  • Parallel [ST] joints
  • Adapter T169500-2440 has parallel [ST] joints
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable