KIMBLE® 105° Distilling Adapters, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
273100-0000 157000-2440 157500-2440 157500-2942
KT273100-0000EA 119.73 CAD
KT273100-0000 KT157000-2440 KT157500-2440 KT157500-2942
KIMBLE® 105° Distilling Adapters, DWK Life Sciences
Distillation Components Distillation Adapters
This distillation bent adapters with tapered drip tube.
  • Standard taper outer joint at top
  • Tapered drip tube extends into open receivers
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.