SC-3 System Easy-On Hoods, WORKLON®
Supplier: Superior Uniform
1050-S 1050-M 1050-L 1050-XL 1050-2XL 1051-S 1051-M 1051-L 1051-XL 1051-2XL
14227-844EA 213.9 CAD
14227-844 14227-846 14227-848 14227-850 14227-852 14227-864 14227-866 14227-868 14227-870 14227-872
SC-3 System Easy-On Hoods, WORKLON®
Workwear
Reusable hoods are designed for use in microelectronics, semiconductor, disk drive, laser, and other industrial ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanrooms
Hoods are made of static dissipative Burlington C3™ material. They drape across the shoulders and feature an adjustable snap front closure, rear horizontal snap adjustment, and two sets of vertical snaps for a facemask attachment. A color-coded hang loop denotes size.
Order Now
SPECIFICATIONS
Garment application Industrial,Cleanroom,Electronics/semi-conductor
Environmentally Preferable
Risk area Anti-static
Gender Unisex
Closure type Press studs / snaps
Fabric Static dissipative Burlington C3™