KIMTECH PREP® SCOTTPURE™ Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
06121 06193
CA21903-010CS 398.5 CAD
CA21903-010 CA89003-700
KIMTECH PREP® SCOTTPURE™ Wipers, KIMBERLY-CLARK PROFESSIONAL®
Wipes Reusable Cleaning Cloths Cloths for General Applications
KIMTECH PREP® SCOTTPURE™ Wipers are designed for wiping tasks where precision and reliability are essential.
Low-lint rayon/polyester spunlace material for cloth-like softness. Engineered for excellent absorbency and low-lint wiping. Solvent resistant. No binders or surfactants. Applications include aerospace, clean manufacturing areas, DVD manufacturing, and printing.