KIMTECH PREP® KIMTEX™ Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
33570 33241 33352 33560
CA21908-646CS 234.85 CAD
CA21908-646 CA21908-668 CA37002-058 CA37002-022
KIMTECH PREP® KIMTEX™ Wipers, KIMBERLY-CLARK PROFESSIONAL®
Wipes Reusable Cleaning Cloths Cloths for General Applications
KIMTECH PREP® KIMTEX™ Wipers feature 100% polypropylene construction for cloth-like feel and durability.
Wipers provide a virtually lint-free cleaning performance and absorb quickly for big cleanups. Engineered for critical applications involving solvents, inks, and paints, they are also perfect for use with lubricants, oil, and grease. Acid-, base-, and solvent-resistant. Available in a variety of formats to meet every need. Color: blue. Applications include furniture finishing, engine assembly/repair, prepackaging wipe down, LCD panel assembly, appliance assembly, printing, and glass or window cleaning.
Order Now
SPECIFICATIONS
Environmentally Preferable