KIMBLE® KONTES® AIRLESS-WARE® Gas Inlet Adapter with Glass Plug, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
211300-1420 211330-1420 211330-2440 211300-2440
KT211300-1420EA 271.43 CAD
KT211300-1420 KT211330-1420 KT211330-2440 KT211300-2440
KIMBLE® KONTES® AIRLESS-WARE® Gas Inlet Adapter with Glass Plug, DWK Life Sciences
Adapters Adapters, Conical Ground Joints
Gas inlet type adapters with inner [ST] joint.
  • Inner [ST] joint
  • Includes glass and PTFE plug
  • Manufactured from 33 expansion, low extractable borosilicate glass
For use with a sleeved stopper.
Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery information: Supplied with KT801001-0002 glass plug and patented pressure/vacuum retainer and KT882311-0000 sleeved stopper.
Order Now
SPECIFICATIONS
Environmentally Preferable