COMPAC5 Compact Multi-Patient Anesthesia Center, VetEquip®
Supplier: Vetequip
901812 901816
89012-504EA 23036.51 CAD
89012-504 97002-250
COMPAC5 Compact Multi-Patient Anesthesia Center, VetEquip®
Animal Supplies Animal Anesthesia Systems
The Compact Multi-Patient Anesthesia center is ideal for high-volume, assembly-line type procedures.
  • Portable models are mounted onto a mobile cart for convenient transport
  • Space is provided within the cart frame for two E-cylinders of oxygen
  • Portable model with shelves features two fold-down shelves, one located on each side of the cart
The evacuation system features two-stage filtration and is available in a benchtop (89012-658) or cabinet (89012-660) style. For exhaust system options, contact your Avantor sales representative. The warm water pump (89012-722) can be used to heat the chambers. It features a large water reservoir, temperature settings from 29.4 to 41.7 °C (85 to 107 °F), and a high flow rate to assure uniform temperature. Non-adjustable O2 regulators (89012-466 and 89012-468) are preset to 50 psi and have DISS male connections.
The center features two independent pediatric Bain breathing/procedure circuits and up to three independent vented induction chambers. All circuits can run at either 0.5 L/min. or 1 L/min. The chambers can be heated with a standard warm water pump. All chambers and circuits are vented, eliminating pollution. No chamber flushing is required. The aluminum unit has a baked enamel finish and features an exhaust pressure gauge, five multi-function switches, and three adjustable induction chamber compartments with three small slide lids, one medium lid, and one large lid with two dividers. The precision, funnel-fill isoflurane vaporizer features temperature and flow rate compensation.
Ordering information: Supplied with a manufacturer’s one-year money-back guarantee and seven-year replacement warranty; “VetEquip Anesthesia Book” and DVD; isoflurane funnel-fill vaporizer; anti-spill adapter for vaporizer; patient circuits; 0-5 L/min. flow verification tube; 3 m (10') O2 hose with DISS fittings; hoses to connect to warm water pump; and T-shirt.