Adapter, Claisen, Ace Glass Incorporated
Supplier: Ace Glass
5055-15 5055-35 5055-10 9067-02 9067-04
89051-256EA 255.29 CAD
89051-256 89051-258 89051-338 89064-818 89064-820
Adapter, Claisen, Ace Glass Incorporated
Distillation Components Distillation Adapters
This Claisen-style adapter features a parallel sidearm.
  • Parallel sidearm
  • Reinforced outer [ST] joints
  • Manufactured from borosilicate glass
Featuring a reinforced [ST] outer joint or spherical socket joint at the top and a parallel side arm with reinforced [ST] outer joint or spherical socket joint. Adapter bottoms feature either [ST] inner joint or spherical ball joint. All full-sized outer [ST] joints are reinforced.