Septum Inlet Adapters with Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
5111-09 9094-04 5111-19 9094-14
89051-328EA 354.31 CAD
89051-328 89064-898 89084-936 89085-086
Septum Inlet Adapters with Stopcock, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
Sampling adapters are designed for use in handling air-sensitive materials.
  • Bottom [ST] joint
  • Manufactured from borosilicate glass
  • Two septa included
Adapters feature a [ST] joint at bottom, 2mm bore stopcock, and two right angle septum ports at top.
Ordering information: Supplied with two septa. For replacement septa, contact your Avantor sales representative.