Gas Supply Regulators and Flowmeters, VetEquip®
Supplier: Vetequip
401720 401721 401722 401726 401728 401730 401735 401740 401745 401746 401748 411722 411723 901302 901303 901304 901305 901306 901307 901308
89012-410EA 284.8 CAD
89012-410 89012-412 89012-414 89012-416 89012-418 89012-420 89012-424 89012-426 89012-428 89012-430 89012-432 89012-434 89012-436 89012-460 89012-462 89012-464 89012-466 89012-468 89012-470 89012-472
Gas Supply Regulators and Flowmeters, VetEquip®
Animal Supplies Animal Anesthesia Systems
Carrier gas can be supplied from a small cylinder or large tank in the procedure room, on an anesthesia cart, or piped through a wall outlet.
  • Gas-specific regulators have a DISS male connection and are preset to 50 psi
  • Flowmeters can be ordered to replace existing units or added to create a custom machine with varying carrier gas combinations
Regulators and flowmeters include a manufacturer’s one-year money-back guarantee and seven-year replacement warranty.