WypAll® X70 Wipers, Kimberly-Clark Professional®
Supplier: Kimberly-Clark
41600 41100 41600 41061 41611 41702 41200 41455 55300
CA21907-032CS 168.6 CAD
CA21907-032 CA37002-052 CA89000-688 CA89003-012 CA89003-018 CA89003-020 CA37002-028 CA89003-016 76325-982
WypAll® X70 Wipers, Kimberly-Clark Professional®
Wipes Reusable Cleaning Cloths Cloths for General Applications
WypAll® X70 Cloths are durable, reusable, absorbent, and stay strong for better cleaning performance.
  • Fast absorbing HYDROKNIT® material is a hydroentangled cellulose/polypropylene blend with no adhesives or chemical binders
  • Durable, rag-tough construction and textured surface
  • Reusable, will not fall apart when wet
  • Ideal for cleaning oil, dirt, grime, and solvents
  • Picks up water almost instantly, and hangs on for fewer drips, outperforming laundered shop rags
For use in heavy manufacturing, transportation fleet maintenance, metal fabrication, machineMore Product Information