Dry Cotton Cleanroom Wipers, Texwipe®
Supplier: Texwipe
TX304 TX306 TX309 TX312
TW-TX304CS 1803.84 CAD
TW-TX304 TW-TX306 TW-TX309 TW-TX312
Dry Cotton Cleanroom Wipers, Texwipe®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These wipers are made from 100% cotton material with a cut edge, cleanroom manufactured.
  • Designed for use on abrasive surfaces
  • Will not easily snag or abrade releasing particles and fibers into the process or environment
  • Bias-cut to minimize edge shedding and fraying
  • Made from sustainable and renewable materials grown and processed in the US
  • Excellent chemical resistance for compatibility with a variety of solutions
  • Autoclave safe
The wipers are constructed from long staple 100% cotton yarn to reduce free floating fibers on the surface while still providing excellent wiping and cleaning. Minimizes cleanroom contamination with low-lint surface and edges protected by our ULP (Ultra-Low Particulate) treatment.
Cleanroom Environment: ISO Class 6 - 8, Class 1,000 - 100,000, EU Grade B - D