DuPont™ ProClean® Frocks, Front Snap Closure
Supplier: DuPont
PC270SWHSM00300B PC270SWHMD00300B PC270SWH3X00300B PC270SWH4X00300B PC270SWH2X00300B
CA13452-502CS 1055.95 CAD
CA13452-502 CA13452-504 CA13452-512 CA13452-514 CA89129-702
DuPont™ ProClean® Frocks, Front Snap Closure
Disposable Garments
ProClean® offers improved non-hazardous light liquid splash protection.
  • Serged seams
  • Bound neck
  • Set sleeve design
  • Covered elastic wrists
  • Front snap closure
ProClean® is made from a microporous composite fabric. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Covered elastic openings at wrists allows for a tighter fit. Bound neck ensures lower particle shedding. Frocks provide body-length jacket design with front snap closures for easy donning and doffing.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners.