Type I Nonmetallic Safety Cans, Justrite®
Supplier: Justrite
14065 14160 14261 14561
56609-592EA 289.78 CAD
56609-592 56609-594 56609-595 56609-621
Type I Nonmetallic Safety Cans, Justrite®
Safety Cans
High-density polyethylene cans are dent-, puncture-, and chip-resistant
  • Inert and resistant to tough acids, corrosives, and heat
  • Feature dual-density flame arrester, self-closing cap, and pressure relief mechanism
  • Current-carrying carbon insert imbedded in the can provides proper grounding
  • Insert prevents the creation of arc during filling or pouring
  • All cans are FM approved
  • Round cans are UL and C-UL listed
They feature a conductive current-carrying carbon insert embedded in the rib of the container, completing a ground path between the cover assembly and the flame arrester in the spout. When used with a grounding strip, this provides proper grounding and prevents the creation of an arc during filling or pouring. The self-closing cap, pressure relief mechanism, and dual-density flame arrester work together to prevent ignition sources without restricting liquid flow in or out of the can.
Cans are inert to resist tough acids and corrosives as well as heat.
Certifications: Cans are available with steel-plated or stainless steel hardware. FM approved. Round cans are UL and C-UL listed.