Bench Cans, Justrite®
Supplier: Justrite
10175 10295 10370 10375 10575 10775
56609-720EA 273.48 CAD
56609-720 56609-722 56609-724 56609-726 56609-728 56609-730
Bench Cans, Justrite®
Safety Cans
Terne plate steel bench can for convenient cleaning of small parts in solvents
Simply place part on spring-loaded, perforated dasher plate, press down and agitate, then release to return dasher to drain position. Plated steel dasher is an effective flame arrester. Hinged cover minimizes evaporation losses. Protected rim reduces possibility of hand injury. Item 56609-724 is a can with parts basket.
Certifications: FM approved.
Order Now
SPECIFICATIONS
Environmentally Preferable