Safety Plunger Cans, Eagle Manufacturing
Supplier: Eagle Mfg
P-711 P-704 P-701 P-712 P-702 P-714
21815-443EA 277.7 CAD
21815-443 21815-421 21815-400 21815-470 21815-450 21815-460
Safety Plunger Cans, Eagle Manufacturing
Safety Cans
For safe cleaning with flammable or volatile liquids.
Close control of liquid is provided by spring-mounted dasher, which forces a measured amount of fluid to rise and dampen swab, sponge, or cloth. No messy or dangerous spills. The surplus liquid drains back into the can to help eliminate waste and hazards. Dasher screen acts as a fire baffle. Plunger assembly is easily removed for cleaning. Dasher diameter: 13.3cm (51/4").
Dent-proof and puncture-resistant polyethylene models are ideal for use with high purity and corrosive chemicals.
Certifications: Complies with OSHA standards. FM approved.