Safety Disposal Cans, Type 316 Stainless Steel, Eagle Manufacturing
Supplier: Eagle Manufacturing
1323 1325
56610-130EA 2721.77 CAD
56610-130 56610-141
Safety Disposal Cans, Type 316 Stainless Steel, Eagle Manufacturing
Safety Cans
Designed for use as an intermediate station for liquid wastes.
  • Designed for use as an intermediate station for liquid wastes
The 10.1cm (4") I.D. opening permits these cans to be filled and emptied easily. Spout cap can be locked open for convenient emptying of many types of containers without use of a funnel. Each can is equipped with a flame arrester to reduce the possibility of ignition of contents by external spark. The cap assembly opens automatically to relieve excessive vapor pressure built up inside the can. The bottom of each can features a reinforced band of heavy-gauge steel for extra strength. Free-swinging handle maintains balance when can is carried, empty or full. With stainless steel fittings.
Certifications: FM approved.