SC-3 System Frocks, WORKLON®
Supplier: Superior Uniform
3441-XS 3441-S 3441-L 3441-XL 3441-2XL 3441-3XL 3441-4XL 3441-5XL 3442-XS 3442-S 3442-M 3442-L 3442-XL 3442-2XL 3442-3XL 3442-4XL 3442-5XL 3440-XS 3440-S 3440-M 3440-L 3440-XL 3440-2XL 3440-3XL 3440-4XL 3440-5XL
14227-976EA 534.85 CAD
14227-976 14227-978 14227-982 14227-984 14227-986 14227-988 14227-990 14227-992 14227-994 14227-996 14227-998 14228-000 14228-002 14228-004 14228-006 14228-008 14228-010 14228-012 14228-014 14228-016 14228-018 14228-020 14228-022 14228-024 14228-026 14228-028
SC-3 System Frocks, WORKLON®
Workwear
Reusable frocks are designed for use in microelectronics, semiconductor, disk drive, laser, and other ISO Class 6 (FED-STD-209E Class 1000/M4.5) industrial cleanrooms.
  • Available with snap front closure or zipper front closure
  • Color-coded hang loop denotes size
The Burlington C3™ high density fabric dissipates static charges and guards against airborne particles and lint. Reusable cleanroom frocks have a two-ply reinforced placket, antistatic knit cuffs, and adjustable snaps at the collar.
Order Now
SPECIFICATIONS
Environmentally Preferable
Garment application Laboratory Laboratory
Risk area Anti-static Anti-static
Gender Unisex Unisex
Sleeve or leg length Long Long
Closure type Press studs / snaps Zip closure