LD-100 System Frocks, 3454 Series, WORKLON®
Supplier: Superior Uniform
3454-XS 3454-S 3454-M 3454-L 3454-XL 3454-2XL 3454-3XL 3454-4XL 3454-5XL
14228-044EA 232.62 CAD
14228-044 14228-046 14228-048 14228-050 14228-052 14228-054 14228-056 14228-058 14228-060
LD-100 System Frocks, 3454 Series, WORKLON®
Workwear
Reusable cleanroom frocks are designed for use in pharmaceutical, medical device, food processing, and other industries where particulate control and bacteria filtration are required.
Frocks are constructed of 100% polyester taffeta with polyester knit cuffs, raglan sleeves, and adjustable snaps at the collar
Suitable for ISO Class 6 (FED-STD-209E Class 1000/M4.5) controlled environments.
A color-coded hang loop denotes size. Frocks have a duo closure style that features a zipper front closure as well as one lower snap. Two-ply reinforced placket. Gamma compatible. Autoclavable.