Silver Shield®/4H® Coat Aprons, Honeywell Safety
Supplier: Honeywell Safety Products
SSCA/XL SSCA/L SSCA/M SSCA/S
CA11000-632CS 1446.59 CAD
CA11000-632 CA11000-634 CA11000-636 CA11000-638
Silver Shield®/4H® Coat Aprons, Honeywell Safety
Disposable Garments
Aprons feature a five-layer fabric compacted into 2.7 mil.
  • Each layer acts as a barrier against various epoxies and industrial chemicals
  • Impervious to at least 90% of all organic chemicals for 4 hours at 35 °C (95 °F)
  • Cut from a single piece of film to prevent penetration of liquid chemicals
  • Sleeves with flexible wrist openings, buckle closure attached to neckband, two waistbands that tie in the back
Each apron weighs less than 198 g (7oz).