Vinyl Coat Aprons, Ansell
Supplier: Ansell Healthcare
950191 950193 950192
32890-884CS 731.75 CAD
32890-884 32890-888 32891-286
Vinyl Coat Aprons, Ansell
Disposable Garments
SType 56-910 aprons provide excellent protection when handling acids and caustic solutions.
  • Heat-sealed binding at the neck and bottom eliminates leaks caused by exposed elastic and thread
  • Back closure consists of two plastic snaps and nylon ties, with no metal parts to cause static or corrosion
  • Full-length sleeves provide additional coverage
Made of virgin vinyl.