Universal Inlet Adapters, Chemglass
Supplier: Chemglass
CG-1042-01 CG-1042-14
80065-972EA 99.94 CAD
80065-972 80065-980
Universal Inlet Adapters, Chemglass
Adapters Adapters, Joints with Tube Connectors
These universal adapters have a thread at the top for a compression cap and o-ring.
  • Universal
  • Threaded top
  • Inner [ST] joint
Adapters feature a #7 em-Thread at the top for compression cap and Viton® O-ring.