TuffStuff™ Critical Task Wipers, Contec®
Supplier: Contec
TSWC0917 TSWC1217 C1C1217Q TSWL1213
33502-722CS 356.99 CAD
33502-722 33502-724 33503-134 94002-798
TuffStuff™ Critical Task Wipers, Contec®
Wipes Disposable Wipes and Towels Wipes for Delicate Tasks
TuffStuff™ Wipes are high-quality critical task wipes. Available in medium and heavy-duty formats, these wipes are solvent resistant and maintain their strength even when wet. TuffStuff™ wipes offer exceptional durability and cleanliness.
  • Hydroentangled nonwoven wipes are strong and durable without the use of binders
  • Medium and heavy-duty wipes available to match the ideal wipe for the specific application
  • Offers strength wet or dry, no residues or binders left behind
  • Highly sorbent without the use of surfactants
  • Multiple wipe sizes allow optimum selection
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable