WorkWipes® Series 60 Heavy-Duty Wipers, New Pig
Supplier: New Pig
WIP661 WIP660 WIP662
89073-946EA 254.3 CAD
89073-946 89073-944 89073-948
WorkWipes® Series 60 Heavy-Duty Wipers, New Pig
Wipes Disposable Wipes and Towels Wipes for General Applications
Lightest of the heavy-duty WorkWipes® series, these economical wipers are made of cellulose and synthetic fibers for strength and low linting properties.
  • Textured surface helps remove dirt and grime
  • Ideal for applying disinfectants, degreasers, and solvents
Additional applications include grease removal, wiping hands, fast cleanups, and cleaning glass or stainless steel.