WYPALL® L40 Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
03046 05007 05740 05790 05776 05796 05701
CA14224-028CS 260.36 CAD
CA14224-028 CA82007-774 CA89000-684 CA89000-692 CA89002-998 CA89011-792 CA21908-000 CA21908-564
WYPALL® L40 Wipers, KIMBERLY-CLARK PROFESSIONAL®
Wipes Disposable Wipes and Towels Wipes for General Applications
A lighter-weight wiper at a great everyday price.
  • 100% virgin fiber
  • Extra low linting, anti-static
  • Wipe clean without scratching or leaving any residue
  • For wiping surfaces, parts, instruments, and lens cleaning
Disposable wipers feature double re-creped (DRC) base sheet technology with high surface softness to absorb large liquid spills and leaks. The reinforced grid provides strength for wet-surface wiping and cleaning. Ideal for routine industrial cleaning and maintenance as well as for removing soils and liquids from face and hands.
Handy, space-More Product Information