KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
34743 34705 34721 34133 34155 34256 34120 34256 34120
CA10805-905CS 247.9 CAD
CA10805-905 CA21903-005 CA21903-021 CA21905-011 CA21905-026 CA21905-049 CA72390-010 21905-049 89218-057
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Wipes Disposable Wipes and Towels Wipes for Delicate Tasks
KIMTECH SCIENCE® KIMWIPES™ delicate task wipers were introduced over 60 years ago, and since then have been the market leader for wiping surfaces, parts, instruments, and lens cleaning in laboratories, research environments, or anywhere delicate wiping is required.
  • For wiping surfaces, parts, instruments, and lens cleaning
  • Low-extractable wipers contain 100% virgin fiber
  • Wipe clean without scratching or leaving any residue
  • Handy, space-saving POP-UP™ dispenser box dispenses wipers one at a time, to reduce waste
  • Anti-static
The creped surface picks up and holds oil, dust, and liquids from glass, plastic, metal, and wood surfaces. Anti-static dispensing reduces lint; electrostatic discharge controls usage.