VWR® Spec-Wipe® 4 Sterile Wiper
Supplier: VWR
VWR® Spec-Wipe®
21914-772
21914-772CS 1116.57 CAD
21914-772
VWR® Spec-Wipe® 4 Sterile Wiper
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
Wiper offers superior quality for aseptic environments where bacteria, particulate, and extractable levels are critical concerns.
  • 100% double-knit polyester
  • Continuous filament
  • Superior quality for aseptic environments
This 100% double-knit polyester wiper has low levels of contaminants as a result of its tightly spun, continuous-filament material and clean-cut edges.
While designed to be durable for rigorous cleaning, it is soft enough for delicate surfaces. Wiper is laundered in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom and sterilized by gamma radiation to a Sterility Assurance Level of 10-⁶ in accordance with ANSI/ISO/AAMI guidelines.
Certifications: Laundered in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
Packaging: 20 wipers per bag. 15 bags per case.