PROSAT® Presaturated Knitted Polynit Wipes, Contec®
Supplier: Contec
PSPS0030 PS-HS-9010 PSPS0002
89347-996CS 1232.73 CAD
89347-996 89368-932 76266-226 76271-056
PROSAT® Presaturated Knitted Polynit Wipes, Contec®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Premoistened polyester wipes are designed for general-purpose cleaning in ISO Class 5 (FED-STD-209E Class 100/M3.5) environments.
  • Ideal for general purpose and critical cleaning
  • Wipes are chemical resistant and exceptionally low in particles and extractable residue
  • Presaturation greatly reduces solvent usage
Standard weight wipes are made of 100% textured polyester with no-run interlock knit for good solvent sorbency, abrasion resistance, and chemical resistance. Heatsealed and saturated with various blends of isopropyl alcohol and deionized water. Low in particles and extractables. Laundered and packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
Sterile wipers are Validated Sterile to 10⁻⁶ Sterility Assurance level in accordance with AAMI/ISO 11137 Method 1 Guidelines for the sterilization of medical devices. Sterile PROSAT® Knitted Polynit Wipes are conveniently packaged for use in sterile core areas and are ideal for wiping articles prior to pass-through and environmental cleaning.
Ideal for wiping exterior product surfaces and cleaning the interior surfaces of process tools or other equipment.
Ordering information: Wipers are packaged in foil pouches or resealable pouches. Resealable pouches allow easy dispensing and help to preserve cleanliness and solvent saturation levels. Pouches are individually lot-numbered for traceability.